Methyl-2-nitrobenzoate ≥98%

Supplier: Thermo Scientific

Synonyms: Methyl 2-nitrobenzoate, 2-Nitrobenzoic acid emthyl ester

B23009.14 B23009.22
B23009.14EA 86 EUR
B23009.14 B23009.22
Methyl-2-nitrobenzoate ≥98%
Methyl 2-nitrobenzoate

Purity: 98+% CAS No.: 606-27-9 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Flash Point: 110°(230°F) Refractive Index: 1.5330 Melting Point: -13° Boiling Point: 275° Beilstein Reference: 2050101 MDL No.: MFCD00007136

Formula: C₈H₇NO₄
MW: 181,15 g/mol
Boiling Pt: 275 °C
Melting Pt: –13 °C
Density: 1,285
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007136
CAS Number: 606-27-9
EINECS: 210-111-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR