2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
L20209.03
L20209.06
L20209.14
L20209.03EA
44.6
EUR
L20209.03
L20209.06
L20209.14
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥97%
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
Purity: 97%
CAS No.: 652-34-6
Molecular Formula: C7H2F4O3
Formula Weight: 210.10
Melting Point: 148-150° dec.
Beilstein Reference: 986011
Formula:
C₇H₂F₄O₃ MW: 210,08 g/mol Melting Pt: 148…150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00129952 CAS Number: 652-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...