2,6-Difluorophenyl isothiocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,6-Difluorophenyl isocyanate, 2,6-Difluorophenylisothiocyanate, 1,3-Difluoro-2-isothiocyanatobenzene
L11903.03
L11903.06
L11903.03EA
43.4
EUR
L11903.03
L11903.06
2,6-Difluorophenyl isothiocyanate ≥97%
2,6-Difluorophenyl isothiocyanate
Purity: 97%
CAS No.: 207974-17-2
Molecular Formula: C7H3F2NS
Formula Weight: 171.17
Flash Point: 104°(219°F)
Refractive Index: 1.6020
Boiling Point: 217°
Beilstein Reference: 2090586
UN No.: 2922
MDL No.: MFCD00041047
Formula:
C₇H₃F₂NS MW: 171,17 g/mol Boiling Pt: 217 °C Density: 1,325 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041047 CAS Number: 207974-17-2 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...