2-(Trifluoromethoxy)benzenesulphonyl chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(Trifluoromethoxy)benzene-1-sulphonyl chloride
A10707.03
A10707.06
A10707.03EA
67.1
EUR
A10707.03
A10707.06
2-(Trifluoromethoxy)benzenesulphonyl chloride ≥97%
2-(Trifluoromethoxy)benzenesulphonyl chloride
Purity: 97%
CAS No.: 103008-51-1
Molecular Formula: C7H4ClF3O3S
Formula Weight: 260.62
Refractive Index: 1.4825
Melting Point: 29-32°
Boiling Point: 75-77°/1mm
Beilstein Reference: 8482182
UN No.: 3261
MDL No.: MFCD00052316
Formula:
C₇H₄ClF₃O₃S MW: 260,62 g/mol Boiling Pt: 75…77 °C (1 mmHg) Melting Pt: 29…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052316 CAS Number: 103008-51-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...