3-(Trifluoromethoxy)benzenesulphonyl chloride 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-(Trifluoromethoxy)benzene-1-sulphonyl chloride
L19552.03
L19552.06
L19552.MD
L19552.03EA
113
EUR
L19552.03
L19552.06
L19552.MD
3-(Trifluoromethoxy)benzenesulphonyl chloride 95%
3-(Trifluoromethoxy)benzenesulphonyl chloride
Purity: 95%
CAS No.: 220227-84-9
Molecular Formula: C7H4ClF3O3S
Formula Weight: 260.62
Refractive Index: 1.4754
Melting Point: 229-230°
Boiling Point: 73-74°/0.7mm
UN No.: 3265
MDL No.: MFCD01091016
Formula:
C₇H₄ClF₃O₃S MW: 260,62 g/mol Boiling Pt: 73…74 °C (0,7 mmHg) Melting Pt: 229…230 °C Density: 1,53 Storage Temperature: Ambient |
MDL Number:
MFCD01091016 CAS Number: 220227-84-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...