Azobenzene 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
401540500
401542500
ACRO401540500EA
140
EUR
ACRO401540500
ACRO401542500
Azobenzene 98%
Azobenzene
Formula:
C₁₂H₁₀N₂ Boiling Pt: 293 °C (1013 hPa) Melting Pt: 66…69 °C Density: 1,203 g/cm³ (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003022 CAS Number: 103-33-3 EINECS: 203-102-5 UN: 3077 ADR: 9,III Merck Index: 12,00948 |
Specification Test Results
Appearance | Orange to red Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 65 to 69 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...