3-Nitrophenyl isocyanate 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Isocyanato-3-nitrobenzene, 3-Nitrophenylisocyanate

336180050 336180250
ACRO336180050EA 61.2 EUR
ACRO336180050 ACRO336180250
3-Nitrophenyl isocyanate 97%
3-Nitrophenyl isocyanate
Formula: C₇H₄N₂O₃
Boiling Pt: 130…131 °C (11 mmHg)
Melting Pt: 50…52 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007220
CAS Number: 3320-87-4
EINECS: 222-025-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR