4-Ethoxyphenyl isocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Ethoxy-4-isocyanatobenzene
L11509.06
L11509.14
L11509.06EA
46.6
EUR
L11509.06
L11509.14
4-Ethoxyphenyl isocyanate ≥97%
4-Ethoxyphenyl isocyanate
Purity: 97%
CAS No.: 32459-62-4
Molecular Formula: C9H9NO2
Formula Weight: 163.18
Storage Temperature: 2°C to 8°C
Flash Point: 105°(221°F)
Refractive Index: 1.5360
Boiling Point: 235°
Beilstein Reference: 742647
UN No.: 2206
MDL No.: MFCD00013877
Formula:
C₉H₉NO₂ MW: 163,18 g/mol Boiling Pt: 235 °C Density: 1,110 Flash Pt: 105 °C (221 °F) |
MDL Number:
MFCD00013877 CAS Number: 32459-62-4 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...