2-Amino-4-nitrophenol 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-5-nitroaniline

A13964.14 A13964.22
A13964.14EA 89.4 EUR
A13964.14 A13964.22
2-Amino-4-nitrophenol 98%
2-Amino-4-nitrophenol

Purity: 98% CAS No.: 99-57-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 141-145° Beilstein Reference: 776533

Formula: C₆H₆N₂O₃
MW: 154,13 g/mol
Melting Pt: 141…145 °C
Density: 1,66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR