2-Aminothiophenol 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A18952.14
A18952.22
A18952.36
A18952.14EA
34.8
EUR
A18952.14
A18952.22
A18952.36
2-Aminothiophenol 98%
2-Aminothiophenol
Purity: 98%
CAS No.: 137-07-5
Molecular Formula: C6H7NS
Formula Weight: 125.19
Flash Point: 79°(174°F)
Refractive Index: 1.6420
Odour: Stench
Melting Point: 18-23°
Boiling Point: 234°
Beilstein Reference: 606076
UN No.: 3267
MDL No.: MFCD00007702
Formula:
C₆H₇NS Boiling Pt: 234 °C Melting Pt: 18…23 °C Density: 1,170 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007702 CAS Number: 137-07-5 EINECS: 205-277-3 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...