2-Nitroresorcinol 98
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Nitrobenzene-1,3-diol
A12970.06
A12970.14
A12970.22
A12970.06EA
47.4
EUR
A12970.06
A12970.14
A12970.22
2-Nitroresorcinol 98
2-Nitroresorcinol
Purity: 98%
CAS No.: 601-89-8
Molecular Formula: C6H5NO4
Formula Weight: 155.11
Melting Point: 81-85°
Beilstein Reference: 2048819
UN No.: 2811
MDL No.: MFCD00007124
Formula:
C₆H₅NO₄ MW: 155.11 g/mol Boiling Pt: 232 Melting Pt: 81…85 °C Density: 1.3526 g/cm³ (96.7 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007124 CAS Number: 601-89-8 EINECS: 210-010-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...