2-Nitrophenyl phenyl ether ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Nitrodiphenylether, 1-Nitro-2-phenoxybenzene, 2-Nitrophenylphenylether
A18088.09
A18088.18
A18088.30
A18088.09EA
43.7
EUR
A18088.09
A18088.18
A18088.30
2-Nitrophenyl phenyl ether ≥99%
2-Nitrophenyl phenyl ether
Purity: 99%
CAS No.: 2216-12-8
Molecular Formula: C12H9NO3
Formula Weight: 215.21
Flash Point: 181°(358°F)
Refractive Index: 1.6 00
Boiling Point: 184°/8mm
Beilstein Reference: 2051914
MDL No.: MFCD00035744
Formula:
C₁₂H₉NO₃ MW: 215,21 g/mol Boiling Pt: 184 °C (8 mmHg) Density: 1,230 Flash Pt: 181 °C (358 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00035744 CAS Number: 2216-12-8 EINECS: 218-684-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...