2-Nitrobenzenesulphonyl chloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-Nitrobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

8.42593.0025 8.42593.0100
8.42593.0025EA 46.4 EUR
8.42593.0025 8.42593.0100
2-Nitrobenzenesulphonyl chloride for synthesis, Sigma-Aldrich®
2-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
MW: 221,62 g/mol
Melting Pt: 69,5…69.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00007430
CAS Number: 1694-92-4
EINECS: 216-907-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 65 - 67 °C
Molar Mass 221.62 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR