2-(Methylthio)phenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxythioanisole

L10128.06 L10128.03 L10128.14
L10128.06EA 133 EUR
L10128.06 L10128.03 L10128.14
2-(Methylthio)phenol ≥97%
2-(Methylthio)phenol

Purity: 97+% CAS No.: 1073-29-6 Molecular Formula: C7H8OS Formula Weight: 140.21 Flash Point: 100°(212°F) Refractive Index: 1.5930 Boiling Point: 104-106°/22mm Beilstein Reference: 1859745 MDL No.: MFCD00002211

Formula: C₇H₈OS
Boiling Pt: 104…106 °C (22 mmHg)
Density: 1,16
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002211
CAS Number: 1073-29-6
EINECS: 214-027-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR