Diphenyl ether ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

A15791.0E A15791.30 A15791.36
A15791.0EEA 105 EUR
A15791.0E A15791.30 A15791.36
Diphenyl ether ≥99%
Diphenyl ether

Purity: 99% CAS No.: 101-84-8 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 125°(257°F) Refractive Index: 1.5805 Odour: Aromatic Melting Point: 26-29° Boiling Point: 258-260° Beilstein Reference: 1364620 Merck Reference: 14,7288 UN No.: 3077 MDL No.: MFCD00003034

Formula: C₁₂H₁₀O
Boiling Pt: 258…260 °C
Melting Pt: 26…29 °C
Density: 1,071
Flash Pt: 125 °C (257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9,III
Merck Index: 14,7288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR