Benzenesulphonyl chloride 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Benzene sulphochloride
148480010
148482500
ACRO148480010EA
138
EUR
ACRO148480010
ACRO148482500
Benzenesulphonyl chloride 99%
Benzenesulphonyl chloride
Formula:
C₆H₅ClO₂S MW: 176,62 g/mol Boiling Pt: 251…252 °C (760 mmHg) Melting Pt: 14,5 °C Density: 1,384 g/cm³ Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007426 CAS Number: 98-09-9 EINECS: 202-636-6 UN: 2225 ADR: 8,III Merck Index: 13,01072 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5530 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...