4-Formylphenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Boronobenzaldehyde, 4-Formylbenzeneboronic acid, (4-Formylphenyl)metaboric acid, Dihydroxy(4-formylphenyl)borane

B25199.03 B25199.06 B25199.14
B25199.03EA 50.4 EUR
B25199.03 B25199.06 B25199.14
4-Formylphenylboronic acid ≥97%
4-Formylphenylboronic acid

Purity: 97% CAS No.: 87199-17-5 Molecular Formula: C7H7BO3 Formula Weight: 149.94 Melting Point: 260-266° Beilstein Reference: 3030770

Formula: C₇H₇BO₃
MW: 149,94 g/mol
Melting Pt: 260…266 °C
Storage Temperature: Ambient
MDL Number: MFCD00151823
CAS Number: 87199-17-5
EINECS: 438-670-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR