1-Bromo-4-butylbenzene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Bromophenyl)butane

B23979.14 B23979.22 B23979.36
B23979.14EA 55.4 EUR
B23979.14 B23979.22 B23979.36
1-Bromo-4-butylbenzene ≥98%
1-Bromo-4-butylbenzene

Purity: 98% CAS No.: 41492-05-1 Molecular Formula: C10H13Br Formula Weight: 213.12 Flash Point: 108°(226°F) Refractive Index: 1.5300 Boiling Point: 242° Beilstein Reference: 2080263 MDL No.: MFCD00040934

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 242 °C
Melting Pt: –26 °C
Density: 1.208
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040934
CAS Number: 41492-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR