2,4,5-Tribromotoluene ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Methyl-2,4,5-tribromobenzene, 1,2,4-Tribromo-5-methylbenzene

A13507.03 A13507.06 A13507.14
A13507.03EA 0 EUR
A13507.03 A13507.06 A13507.14
2,4,5-Tribromotoluene ≥97%
2,4,5-Tribromotoluene

Purity: 97+% CAS No.: 3278-88-4 Molecular Formula: C7H5Br3 Formula Weight: 328.83 Odour: Odourless Melting Point: 112-115° MDL No.: MFCD00052984

Formula: C₇H₅Br₃
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00052984
CAS Number: 3278-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR