α-Bromo-5-chloro-2-fluorotoluene ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Chloro-2-fluorobenzyl bromide, 2-(Bromomethyl)-4-chloro-1-fluorobenzene

L19147.03 L19147.MD
L19147.03EA 95.7 EUR
L19147.03 L19147.MD
α-Bromo-5-chloro-2-fluorotoluene ≥97%
α-Bromo-5-chloro-2-fluorotoluene

Purity: 97% CAS No.: 71916-91-1 Molecular Formula: C7H5BrClF Formula Weight: 223.48 Refractive Index: 1.5650 Boiling Point: 44-45°/0.5mm UN No.: 3265 MDL No.: MFCD01631437

Formula: C₇H₅BrClF
MW: 223,47 g/mol
Boiling Pt: 44…45 °C (0,5 mmHg)
Density: 1,65
Storage Temperature: Ambient
MDL Number: MFCD01631437
CAS Number: 71916-91-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR