ω-Bromoacetophenone ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone

A15576.22 A15576.14 A15576.36
A15576.22EA 81.6 EUR
A15576.22 A15576.14 A15576.36
ω-Bromoacetophenone ≥98%
ω-Bromoacetophenone

Purity: 98% CAS No.: 70-11-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odour: Pungent Melting Point: 48-51° Boiling Point: 140°/11mm Beilstein Reference: 606474 Merck Reference: 14,1402 UN No.: 2645 MDL No.: MFCD00000195

Formula: (C₆H₅)C(O)CH₂Br
MW: 199,05 g/mol
Boiling Pt: 140 °C (11 mmHg)
Melting Pt: 48…51 °C
Density: 1,650
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000195
CAS Number: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 14,1402

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR