α-Bromo-2-(difluoromethoxy)toluene ≥97%

Supplier: Thermo Fisher Scientific
Danger

B20931.09 B20931.18 B20931.88
B20931.09EA 0 EUR
B20931.09 B20931.18 B20931.88
α-Bromo-2-(difluoromethoxy)toluene ≥97%
α-Bromo-2-(difluoromethoxy)toluene

Purity: 97% CAS No.: 85684-64-6 Molecular Formula: C8H7BrF2O Formula Weight: 237.04 Flash Point: 108°(228°F) Refractive Index: 1.5180 Boiling Point: 222° UN No.: 3265 MDL No.: MFCD00190103

Formula: C₈H₇BrF₂O
Boiling Pt: 222 °C
Melting Pt: 13 °C
Density: 1,555
Flash Pt: 108 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190103
CAS Number: 85684-64-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR