3',4'-Dichloropropiophenone ≥98%

Supplier: Thermo Fisher Scientific
Warning

A18740.06 A18740.14
A18740.06EA 54.2 EUR
A18740.06 A18740.14
3',4'-Dichloropropiophenone ≥98%
3',4'-Dichloropropiophenone

Purity: 98% CAS No.: 6582-42-9 Molecular Formula: C9H8Cl2O Formula Weight: 203.07 Flash Point: 110°(230°F) Melting Point: 44-46° Boiling Point: 136-140°/18mm MDL No.: MFCD00000554

Formula: C₉H₈Cl₂O
Boiling Pt: 136…140 °C (18 mmHg)
Melting Pt: 44…46 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000554
CAS Number: 6582-42-9
EINECS: 229-511-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR