4-Chloro-m-toluic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-3-methylbenzoic acid

L08533.09 L08533.04 L08533.18
L08533.09EA 97 EUR
L08533.09 L08533.04 L08533.18
4-Chloro-m-toluic acid ≥98%
4-Chloro-m-toluic acid

Purity: 98% CAS No.: 7697-29-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 212-216° Beilstein Reference: 1936535

Formula: C₈H₇ClO₂
MW: 170,6 g/mol
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00045853
CAS Number: 7697-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR