2-Chloro-6-fluorobenzyl alcohol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Chloro-6-fluorophenyl)methanol

A11978.06 A11978.14
A11978.06EA 29.5 EUR
A11978.06 A11978.14
2-Chloro-6-fluorobenzyl alcohol ≥97%
2-Chloro-6-fluorobenzyl alcohol

Purity: 97% CAS No.: 56456-50-9 Molecular Formula: C7H6ClFO Formula Weight: 160.58 Flash Point: 110°(230°F) Melting Point: 42-44° Beilstein Reference: 1938353 MDL No.: MFCD00004608

Formula: C₇H₆ClFO
MW: 160,58 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004608
CAS Number: 56456-50-9
EINECS: 260-192-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR