4-Chloro-m-cresol ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

A17823.22 A17823.36
A17823.22EA 30.4 EUR
A17823.22 A17823.36
4-Chloro-m-cresol ≥99%
4-Chloro-m-cresol

Purity: 99% CAS No.: 59-50-7 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 118°(244°F) Melting Point: 64-68° Boiling Point: 234-236° Beilstein Reference: 1237629 Merck Reference: 14,2133 UN No.: 3437 MDL No.: MFCD00002323

Formula: C₇H₇ClO
Boiling Pt: 234…236 °C
Melting Pt: 64…68 °C
Density: 1,370
Flash Pt: 118 °C (244 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II
Merck Index: 14,2133

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR