2',4',6'-Trimethoxyacetophenone ≥98%
Supplier: Thermo Fisher Scientific
A14328.06
A14328.14
A14328.06EA
65.1
EUR
A14328.06
A14328.14
2',4',6'-Trimethoxyacetophenone ≥98%
2',4',6'-Trimethoxyacetophenone
Purity: 98%
CAS No.: 832-58-6
Molecular Formula: C11H14O4
Formula Weight: 210.23
Melting Point: 101-104°
Beilstein Reference: 519324
Formula:
C₁₁H₁₄O₄ Melting Pt: 101…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017238 CAS Number: 832-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...