3',4'-Dihydroxypropiophenone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13444.03
A13444.06
A13444.03EA
43.5
EUR
A13444.03
A13444.06
3',4'-Dihydroxypropiophenone ≥98%
3',4'-Dihydroxypropiophenone
Purity: 98%
CAS No.: 7451-98-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 145-146°
MDL No.: MFCD00016440
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 145…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016440 CAS Number: 7451-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...