2',5'-Dihydroxyacetophenone ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
A12185.06
A12185.14
A12185.22
A12185.06EA
41.5
EUR
A12185.06
A12185.14
A12185.22
2',5'-Dihydroxyacetophenone ≥98%
2',5'-Dihydroxyacetophenone
Purity: 98+%
CAS No.: 490-78-8
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 204-208°
Beilstein Reference: 637903
Formula:
C₈H₈O₃ MW: 152,15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 204…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002343 CAS Number: 490-78-8 EINECS: 207-716-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...