4-Pentylbenzoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10248.06 A10248.14 A10248.22
A10248.06EA 62.3 EUR
A10248.06 A10248.14 A10248.22
4-Pentylbenzoyl chloride ≥98%
4-Pentylbenzoyl chloride

Purity: 98% CAS No.: 49763-65-7 Molecular Formula: C12H15ClO Formula Weight: 210.71 Flash Point: 110°(230°F) Refractive Index: 1.5300 Boiling Point: 144-145°/10mm Beilstein Reference: 2207887 UN No.: 3265 MDL No.: MFCD00000700

Formula: C₁₂H₁₅ClO
Boiling Pt: 144…145 °C (10 mmHg)
Density: 1,034
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000700
CAS Number: 49763-65-7
EINECS: 256-478-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR