2-Methyl-m-anisic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Methoxy-2-methylbenzoic acid

L18729.18 L18729.09
L18729.18EA 180 EUR
L18729.18 L18729.09
2-Methyl-m-anisic acid ≥97%
2-Methyl-m-anisic acid

Purity: 97% CAS No.: 55289-06-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Beilstein Reference: 510395 MDL No.: MFCD02094039

Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094039
CAS Number: 55289-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR