α,α,α,3,4-Pentafluorotoluene ≥97%
|
Warning
|
Synonyms:
3,4-Difluorobenzotrifluoride, 1,2-Difluoro-4-(trifluoromethyl)benzene
B24096.03
B24096.06
B24096.03EA
51.7
EUR
B24096.03
B24096.06
α,α,α,3,4-Pentafluorotoluene ≥97%
α,α,α,3,4-Pentafluorotoluene
Purity: 97%
CAS No.: 32137-19-2
Molecular Formula: C7H3F5
Formula Weight: 182.10
Refractive Index: 1.3915
Boiling Point: 103-104°
UN No.: 1993
MDL No.: MFCD00077510
Formula:
C₇H₃F₅ MW: 182,09 g/mol Boiling Pt: 103…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077510 CAS Number: 32137-19-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...