3',4'-Difluoropropiophenone ≥97%

Supplier: Thermo Scientific

Synonyms: 1-(3,4-Difluorophenyl)propan-1-one

A17257.06
A17257.06EA 119 EUR
A17257.06
3',4'-Difluoropropiophenone ≥97%
3',4'-Difluoropropiophenone

Purity: 97% CAS No.: 23384-72-7 Molecular Formula: C9H8F2O Formula Weight: 170.16 Refractive Index: 1.4880 Boiling Point: 64-65°/1.8mm MDL No.: MFCD00015505

Formula: C₉H₈F₂O
MW: 170,16 g/mol
Boiling Pt: 64…65 °C (1,8 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00015505
CAS Number: 23384-72-7
EINECS: 245-627-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR