2',5'-Difluoroacetophenone ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(2,5-Difluorophenyl)ethanone, 1-(2,5-Difluorophenyl)ethan-1-one
A13911.06
A13911.14
A13911.06EA
0
EUR
A13911.06
A13911.14
2',5'-Difluoroacetophenone ≥98%
2',5'-Difluoroacetophenone
Purity: 98%
CAS No.: 1979-36-8
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Flash Point: 70°(158°F)
Refractive Index: 1.4882
Odour: Characteristic
Boiling Point: 190-191°
Beilstein Reference: 2963573
MDL No.: MFCD00009898
Formula:
C₈H₆F₂O MW: 156,13 g/mol Boiling Pt: 190…191 °C Density: 1,233 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009898 CAS Number: 1979-36-8 EINECS: 217-837-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...