4'-Fluoroacetophenone ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone

A13950.14 A13950.22 A13950.36
A13950.14EA 33.6 EUR
A13950.14 A13950.22 A13950.36
4'-Fluoroacetophenone ≥99%
4'-Fluoroacetophenone

Purity: 99% CAS No.: 403-42-9 Molecular Formula: C8H7FO Formula Weight: 138.14 Flash Point: 71°(159°F) Refractive Index: 1.5110 Melting Point: 4° Boiling Point: 194-196° Beilstein Reference: 386013 MDL No.: MFCD00000354

Formula: C₈H₇FO
MW: 138,14 g/mol
Boiling Pt: 194…196 °C
Melting Pt: 4 °C
Density: 1,141
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000354
CAS Number: 403-42-9
EINECS: 206-960-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR