2'-Fluoropropiophenone ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: o-Fluoropropiophenone

L12423.09 L12423.04 L12423.18
L12423.09EA 74.7 EUR
L12423.09 L12423.04 L12423.18
2'-Fluoropropiophenone ≥99%
2'-Fluoropropiophenone

Purity: 99% CAS No.: 446-22-0 Molecular Formula: C9H9FO Formula Weight: 152.17 Flash Point: 79°(174°F) Refractive Index: 1.5045 Boiling Point: 89-91°/12mm Beilstein Reference: 3235723 MDL No.: MFCD00009893

Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 89…91 °C (12 mmHg)
Density: 1.102
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009893
CAS Number: 446-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR