2,3-Difluoro-4-toluoyl chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,3-Difluoro-4-methylbenzene-1-carbonyl chloride, 2,3-Difluoro-4-methylbenzoyl chloride
L18170.06
L18170.03
L18170.06EA
0
EUR
L18170.06
L18170.03
2,3-Difluoro-4-toluoyl chloride ≥97%
2,3-Difluoro-4-toluoyl chloride
Purity: 97%
CAS No.: 261763-38-6
Molecular Formula: C8H5ClF2O
Formula Weight: 190.58
Refractive Index: 1.5220
Formula:
C₈H₅ClF₂O MW: 190,58 g/mol Boiling Pt: 211 °C (1013 hPa) Melting Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631645 CAS Number: 261763-38-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...