α,α,α,2-Tetrafluoro-m-toluic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B20280.03
B20280.06
B20280.03EA
25.4
EUR
B20280.03
B20280.06
α,α,α,2-Tetrafluoro-m-toluic acid ≥98%
α,α,α,2-Tetrafluoro-m-toluic acid
Purity: 98%
CAS No.: 115029-22-6
Molecular Formula: C8H4F4O2
Formula Weight: 208.11
Melting Point: 126-128°
MDL No.: MFCD00040980
Formula:
C₈H₄F₄O₂ MW: 208,11 g/mol Melting Pt: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040980 CAS Number: 115029-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...