α,α,α,6-Tetrafluoro-m-toluic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Fluoro-5-(trifluoromethyl)benzoic acid
B20293.03
B20293.06
B20293.14
B20293.03EA
40.3
EUR
B20293.03
B20293.06
B20293.14
α,α,α,6-Tetrafluoro-m-toluic acid ≥98%
α,α,α,6-Tetrafluoro-m-toluic acid
Purity: 98+%
CAS No.: 115029-23-7
Molecular Formula: C8H4F4O2
Formula Weight: 208.11
Melting Point: 101-102°
Beilstein Reference: 5813056
Formula:
C₈H₄F₄O₂ MW: 208,11 g/mol Melting Pt: 101…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061292 CAS Number: 115029-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...