α,α,α-Trifluoro-m-toluic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(Trifluoromethyl)benzoic acid

A16737.09 A16737.18
A16737.09EA 58 EUR
A16737.09 A16737.18
α,α,α-Trifluoro-m-toluic acid ≥98%
α,α,α-Trifluoro-m-toluic acid

Purity: 98+% CAS No.: 454-92-2 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Melting Point: 103-107° Boiling Point: 237-240° MDL No.: MFCD00002519

Formula: C₈H₅F₃O₂
MW: 190,12 g/mol
Boiling Pt: 237…240 °C
Melting Pt: 103…107 °C
Density: 1,371 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002519
CAS Number: 454-92-2
EINECS: 207-230-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR