5-Fluoro-o-toluic acid ≥99%
Supplier: Thermo Scientific
|
Warning
|
A10476.03
A10476.06
A10476.14
A10476.03EA
33.5
EUR
A10476.03
A10476.06
A10476.14
5-Fluoro-o-toluic acid ≥99%
5-Fluoro-o-toluic acid
Purity: 99%
CAS No.: 33184-16-6
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Melting Point: 130-132°
Beilstein Reference: 2249813
Formula:
C₈H₇FO₂ MW: 154,14 g/mol Boiling Pt: 263 °C (1013 hPa) Melting Pt: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042294 CAS Number: 33184-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...