2-Fluorobenzylamine 96%

Supplier: Thermo Scientific
Danger

171940010 171940100 171940500
ACRO171940010EA 27.5 EUR
ACRO171940010 ACRO171940100 ACRO171940500
2-Fluorobenzylamine 96%
2-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125,15 g/mol
Boiling Pt: 73…75 °C (13 mmHg)
Density: 1,09 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5190 (20°C, 589 nm)
GC ≥95 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR