2-Fluorobenzylamine 97%
Supplier: Thermo Scientific
|
Danger
|
A15623.06
A15623.09
A15623.18
A15623.06EA
54.2
EUR
A15623.06
A15623.09
A15623.18
2-Fluorobenzylamine 97%
2-Fluorobenzylamine
Purity: 97%
CAS No.: 89-99-6
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 67°(152°F)
Refractive Index: 1.5170
Odour: Amine-like
Boiling Point: 80-82°/18mm
Beilstein Reference: 508109
UN No.: 2735
MDL No.: MFCD00008107
Formula:
C₇H₈FN MW: 125,15 g/mol Boiling Pt: 80…82 °C (18 mmHg) Density: 1,099 Flash Pt: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008107 CAS Number: 89-99-6 EINECS: 201-957-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...