3-Fluorobenzylamine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(3-Fluorophenyl)methanamine
B23527.06
B23527.09
B23527.14
B23527.06EA
74.2
EUR
B23527.06
B23527.09
B23527.14
3-Fluorobenzylamine 97%
3-Fluorobenzylamine
Purity: 97%
CAS No.: 100-82-3
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 71°(159°F)
Refractive Index: 1.5150
Odour: Amine-like
Boiling Point: 186°
Beilstein Reference: 1446928
UN No.: 2735
MDL No.: MFCD00008113
Formula:
C₇H₈FN MW: 125,15 g/mol Boiling Pt: 186 °C Density: 1,097 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008113 CAS Number: 100-82-3 EINECS: 202-891-3 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...