5-Fluoro-o-toluidine ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Amino-4-fluorotoluene, 5-Fluoro-2-methylaniline

B25381.09 B25381.18
B25381.09EA 32.5 EUR
B25381.09 B25381.18
5-Fluoro-o-toluidine ≥98%
5-Fluoro-o-toluidine

Purity: 98% CAS No.: 367-29-3 Molecular Formula: C7H8FN Formula Weight: 125.15 Flash Point: 90°(194°F) Refractive Index: 1.5380 Odour: Characteristic Melting Point: 35-38° Boiling Point: 98-100°/15mm Beilstein Reference: 2637584 UN No.: 2811 MDL No.: MFCD00007764

Formula: C₇H₈FN
Boiling Pt: 98…100 °C (15 mmHg)
Melting Pt: 35…38 °C
Density: 1,13 g/cm³ (20 °C)
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007764
CAS Number: 367-29-3
EINECS: 206-689-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR