4-Ethylphenyl isocyanate ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Ethyl-4-isocyanatobenzene

L12718.06 L12718.03 L12718.14
L12718.06EA 57.7 EUR
L12718.06 L12718.03 L12718.14
4-Ethylphenyl isocyanate ≥98%
4-Ethylphenyl isocyanate

Purity: 98% CAS No.: 23138-50-3 Molecular Formula: C9H9NO Formula Weight: 147.18 Storage Temperature: 2°C to 8°C Flash Point: 78°(172°F) Refractive Index: 1.5270 Odour: Pungent Boiling Point: 67-68°/1mm Beilstein Reference: 971100 UN No.: 2206 MDL No.: MFCD00013881

Formula: C₉H₉NO
MW: 147,18 g/mol
Boiling Pt: 67…68 °C (1 mmHg)
Density: 1,024
Flash Pt: 78 °C (172 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013881
CAS Number: 23138-50-3
EINECS: 245-446-9
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR