o-Tolyl isocyanate ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Methylphenyl isocyanate, 1-Isocyanato-2-methylbenzene

L12247.06 L12247.14
L12247.06EA 42.2 EUR
L12247.06 L12247.14
o-Tolyl isocyanate ≥98%
o-Tolyl isocyanate

Purity: 98% CAS No.: 614-68-6 Molecular Formula: C8H7NO Formula Weight: 133.15 Storage Temperature: 2°C to 8°C Flash Point: 92°(197°F) Refractive Index: 1.5345 Odour: Pungent Boiling Point: 186° Beilstein Reference: 507985 UN No.: 2206 MDL No.: MFCD00002009

Formula: C₈H₇NO
MW: 133,15 g/mol
Boiling Pt: 186 °C
Density: 1,074
Flash Pt: 92 °C (197 °F)
MDL Number: MFCD00002009
CAS Number: 614-68-6
EINECS: 210-389-0
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR