4-Octylaniline ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 4-n-Octylaniline

H26713.03 H26713.06
H26713.03EA 63.7 EUR
H26713.03 H26713.06
4-Octylaniline ≥99%
4-Octylaniline

Purity: 99% CAS No.: 16245-79-7 Molecular Formula: C14H23N Formula Weight: 205.34 Flash Point: 110°(230°F) Refractive Index: 1.516 Boiling Point: 175°/13mm MDL No.: MFCD00007929

Formula: C₁₄H₂₃N
MW: 205,34 g/mol
Boiling Pt: 175 °C (13 mmHg)
Melting Pt: 20 °C
Density: 0,898
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007929
CAS Number: 16245-79-7
EINECS: 240-358-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR