4-Nitro-m-toluidine ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Methyl-4-nitroaniline, 4-Nitro-3-methylaniline, 5-Amino-2-nitrotoluene
L13112.06
L13112.14
L13112.06EA
91
EUR
L13112.06
L13112.14
4-Nitro-m-toluidine ≥99%
4-Nitro-m-toluidine
Purity: 99%
CAS No.: 611-05-2
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Odour: Characteristic
Melting Point: 133-137°
Beilstein Reference: 2209112
UN No.: 2660
MDL No.: MFCD00091833
Formula:
C₇H₈N₂O₂ MW: 152,15 g/mol Melting Pt: 133…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00091833 CAS Number: 611-05-2 EINECS: 210-247-8 UN: 2660 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...