2-Methoxybenzylamine 98%

Supplier: Thermo Scientific
Danger

Synonyms: (2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine

170540100 170540500
ACRO170540100EA 46 EUR
ACRO170540100 ACRO170540500
2-Methoxybenzylamine 98%
2-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137,18 g/mol
Boiling Pt: 231…233 °C (760 mmHg)
Density: 1,05 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS Number: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5475 to 1.5495 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR